missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Yeast GLRX1 Yeast Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Gene ID (Entrez) | 850322 |
| Name | GLRX1 Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Gene Symbol | GRX1 |
| Product Type | Recombinant Protein |
| Cross Reactivity | Yeast |
| Species | E. coli |
| Protein Tag | His |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction