missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zebrafish d2hgdh, mitochondrial Protein

A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) d2hgdh, mitochondrial, was expressed in the hosts and tags indicated.

€ 309.00 - € 2645.00

Specifications

Accession Number NP_001074066.1
Gene ID (Entrez) 565889
Name d2hgdh, mitochondrial Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16038780
View Documents Promotion Details
enQuireBio™
QP7418-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16078780
View Documents Promotion Details
enQuireBio™
QP7418-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16028780
View Documents Promotion Details
enQuireBio™
QP7418-EC-100UG
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16058780
View Documents Promotion Details
enQuireBio™
QP7418-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16068780
View Documents Promotion Details
enQuireBio™
QP7418-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16048780
View Documents Promotion Details
enQuireBio™
QP7418-EC-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001074066.1
d2hgdh, mitochondrial Protein
Not determined.
Full Length
Zebrafish
His-SUMO
Tris-based buffer, 50% glycerol
565889
Research Use Only
d2hgdh
Recombinant Protein
E. coli
ANPANPSAAP PRLPFSRVTQ EDLSFFRALL PGRTITDPDL LKSSNVDWLK TVQGSSDVLL RPKTTEGVSQ ILRYCNERNL AVCPQGGNTG LVGGSVPVFD EIILSTSLMN QVFAFDNISG ILTCQAGCVL ENLSHYLEER DFIMPLDLGA KGSCHIGGNV STNAGGLRLL RYGSLRGTVL GLEVVLADGH VLNCLATLRK DNTGYDLKQL FIGSEGTLGV ITAVSILCPR KPKAVNVAFL GCSSFQQLLE TFQCCRGMLG EILSAFEFLD ASCMNLLEKH LKLTNPITEC PFYIVIETAG SNATHDEEKL HQFLEEVMTS SLVTDGTVAT EATKIKALWS LRERVTEALT HEGYTYKYDI SLPVEKIYDL VQDMRRHLGG MAKNVVGYGH VGDGNLHLNI TSPSKDFDLL AAIEPYVYEW TSQWKGSISA EHGLGLKKRN YIYYSKPSEA VALMGSIKAM LDPKGILNPY KTLPDNIN
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications