missing translation for 'onlineSavingsMsg'
Learn More

Recoverin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00 - € 539.00

Specifications

Antigen Recoverin
Dilution Immunohistochemistry-Paraffin 1:5000 - 1:10000
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18258381
View Documents
Novus Biologicals
NBP2-58802
100 μL
€ 539.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18648196
View Documents
Novus Biologicals
NBP2-58802-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recoverin Polyclonal specifically detects Recoverin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Recoverin
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
5957
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SIYAKFFPDTDPKAYAQHVFRSFDSNLDGTLDFKEYVIALHMTTAGKTNQKLEWAFSLYDVDGNGTISKNEVLEIVMAIFKMITPEDVKLLPDDENTPEKRAEKIWKYFGKNDDDKLTEKEFIEGTLANKEILRLIQFEPQKVKEKMKN
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry-Paraffin 1:5000 - 1:10000
Polyclonal
Rabbit
Neuroscience, Vision
cancer associated retinopathy antigen, Protein CAR, RCV1Cancer-associated retinopathy protein, recoverin
RCVRN
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.