Learn More
Manutan Recovery tray wood Combi-Flip, width: 1000 mm, depth: 700 mm
Brand:  Manutan A010516
Encompass_Preferred
Recovery tray wood Combi-Flip, width: 1000 mm, depth: 700 mm
Product Code. 15544197
  • € 11.07 / Pack of 1
Only null left
Estimated Shipment: 27-05-2026