Learn More
OHAUS™ Rectangle Support Stand Base
Product Code. p-7158969 Shop All Ohaus Products
Click to view available options
Dimensions (L x W):
4 x 6 in.
5 x 8 in.
6 x 9 in.
8 x 10 in.
Unit Size:
Each
Product Code. 15732409
Brand: OHAUS™ 30392359
Only null left
Add to basket
Add to basket
OHAUS™ Rectangle Support Stand Base provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Material Cast Iron
Dimensions (L x W) 4 x 6 in.
Length (English) 4 in.
Length (Metric) 102 mm
Width (English) 6 in.
Width (Metric) 152 mm
Product Type Support Stand Base