missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rectum Matched Tumor and Normal Tissue Slides (Tumor)- Frozen

Product Code. 18674701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 Slides
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18674701 10 Slides Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18674701 Supplier Novus Biologicals™ Supplier No. NBP277674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tissue Slides

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 Slides
For Use With (Application) Immunohistochemistry (IHC), Immunohistochemistry (Frozen), ISH, Hematoxylin and Eosin Stain
Species Human
Target Specificity Rectum
Content And Storage Store at -70°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.