Learn More
Witeg™ Borosilicate Glass Laboratory Bottles with Yellow Screw Cap and Graduation
Pour, store, and mix with these Witeg™ Borosilicate Glass Laboratory Bottles with Yellow Screw Cap and Graduation, an essential piece of glassware for your lab.
Brand:  Witeg™ 5526100GE
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11887760
  • € 63.10 / Pack of 10
Estimated Shipment: 08-07-2021
Add to basket
Description
Description
  • Glass
  • Yellow
Specifications
Specifications
ISO 4796
Yes
Yellow
Borosilicate Glass
10Pack
Yellow
GL 45
100 mL
Screw cap and pouring ring
10
Laboratory Bottle