Learn More
Socorex™ Red Capillary Tubes
Specifications
Specifications
| Volume (Metric) | 100 to 200 μL |
| Color | Red |
| Quantity | 100/Pk. |
| Product Type | Capillary Tube |
| For Use With (Equipment) | Acura 846 Positive Displacement Pipette |
| Material | Glass |