Learn More
Kautex™ Red Polypropylene Screw Closures
Product Code. 17828157
Click to view available options
For Use With (Equipment):
10 to 1000 mL Bottle
100 mL Bottle
1500/2500/4000 mL Bottle
250/350 mL Bottle
2500 mL Bottle
50 mL Bottle
500/750/1000 mL Bottle
Diameter (Metric):
28 mm
32 mm
45 mm
54 mm
80 mm
Unit Size:
Each
Pack of 100
Pack of 210
Product Code. 17828157
Brand: Kautex™ 2000083762
Only null left
Add to basket
Add to basket
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
  • PP
  • Red
  • Tamper evident ring
  • PE-foam insert
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Includes Tamper Evident Ring and Polyethylene Foam Insert
Material Polypropylene
Liner Material Polyethylene Foam Insert
Product Type Screw Cap
For Use With (Equipment) 1500/2500/4000 mL Bottle
Diameter (Metric) 80 mm