Learn More
Kartell™ Red Rubber Conical Stoppers
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0113300
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12697775
  • € 14.05 / Pack of 25
Estimated Shipment
09-07-2020
Add to basket
Description
Description
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height
Specifications
Specifications
11mm
General purpose laboratory
Conical Stopper
Conical
Red
15mm
18mm
25/Pk.
11 x 18mm
Rubber