Learn More
Redox Standard 300mV, Reagecon™
BIOPHARMA
Product Code. 11813570 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 11813570
Brand: Reagecon RS300