Learn More
Fisherbrand Redox solution 220mV, 500ml
Brand: Fisherbrand
Redox solution 220mV, 500ml
Additional Details : Weight : 0.54700kg
Brand: Fisherbrand
Redox solution 220mV, 500ml
Additional Details : Weight : 0.54700kg