Learn More
Buerkle™ Reduced Socket with Inner Thread
Manufactured from polypropylene.
Brand: Buerkle™ 8502-3410
7.95 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.03050kg
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Socket Reduced Inner Thread | |
Autoclavable | |
3/4 to 1 in. |
EU Foodstuffs | |
Polypropylene | |
23 mm |