Learn More
Buerkle™ Reduced Socket with Inner Thread
Manufactured from polypropylene.
€ 7.10 - € 8.90
Specifications
Product Type Socket Reduced Inner Thread
Certifications/Compliance EU Foodstuffs
Autoclavable Autoclavable
Material Polypropylene
View More Specs
Products 2
Product Code Brand Width (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Price Quantity & Availability  
11876433
View Documents Promotion Details
Buerkle™
8502-1234
17.5 mm
€ 7.10
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
11886433
View Documents Promotion Details
Buerkle™
8502-3410
23 mm
€ 8.90
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
Description
Description
  • Polypropylene construction
  • Autoclavable to 121°C
  • Safe for use with foodstuffs
  • Supplied in packs of 10
Specifications
Specifications
Socket Reduced Inner Thread
EU Foodstuffs