missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94227-0.02ml
This item is not returnable.
View return policy
Description
REEP1 Polyclonal antibody specifically detects REEP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| REEP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| C2orf23, chromosome 2 open reading frame 23, receptor accessory protein 1, receptor expression-enhancing protein 1, SPG31FLJ13110 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-201 of human REEP1 (NP_075063.1). KEIDDCLVQAKDRSYDALVHFGKRGLNVAATAAVMAASKGQGALSERLRSFSMQDLTTIRGDGAPAPSGPPPPGSGRASGKHGQPKMSRSASESASSSGTA | |
| 0.02 mL | |
| Neurodegeneration, Neuroscience | |
| 65055 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction