missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP2 Antibody (OTI2C7), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73849MFV450
This item is not returnable.
View return policy
Description
REEP2 Monoclonal antibody specifically detects REEP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| REEP2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI2C7 | |
| Flow Cytometry | |
| C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2 | |
| Full length human recombinant protein of human REEP2(NP_057690) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 51308 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction