Learn More
REEP2 Antibody (S07-6C8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19760-50ul
This item is not returnable.
View return policy
Description
REEP2 Monoclonal antibody specifically detects REEP2 in Human samples. It is validated for Western Blot
Specifications
| REEP2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S07-6C8 | |
| Unconjugated | |
| C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2 | |
| A synthetic peptide of human REEP2 (Uniprot # Q9BRK0) | |
| 50 μg | |
| Signal Transduction | |
| 51308 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |