Learn More
REEP2 Antibody (S07-6C8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19760-100ul
 View more versions of this product
Product Code. 18657813
  • € 645.00 / 100µL
Description
REEP2 Monoclonal antibody specifically detects REEP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
REEP2
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
S07-6C8
Unconjugated
C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2
A synthetic peptide of human REEP2 (Uniprot # Q9BRK0)
100 μg
Signal Transduction
51308
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.