missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP5 Antibody (OTI4D2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73850
This item is not returnable.
View return policy
Description
REEP5 Monoclonal specifically detects REEP5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| REEP5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready | |
| C5orf18polyposis coli region hypothetical protein DP1, chromosome 5 open reading frame 18, D5S346deleted in polyposis 1, DP1TB2YOP1, MGC70440, Polyposis locus protein 1, Protein TB2, receptor accessory protein 5, receptor expression enhancing protein 5, receptor expression-enhancing protein 5 | |
| Mouse | |
| 21.3 kDa | |
| 100 μg | |
| 7905 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| OTI4D2 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| REEP5 | |
| Full length human recombinant protein of human REEP5(NP_005660) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction