missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP5 Antibody (OTI4D2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73850PCP
This item is not returnable.
View return policy
Description
REEP5 Monoclonal specifically detects REEP5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| REEP5 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| REEP5 | |
| Full length recombinant protein of human REEP5 (NP_005660) produced in HEK293T cells. | |
| 0.1 mL | |
| 7905.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4D2 | |
| Flow Cytometry | |
| C5orf18polyposis coli region hypothetical protein DP1, chromosome 5 open reading frame 18, D5S346deleted in polyposis 1, DP1TB2YOP1, MGC70440, Polyposis locus protein 1, Protein TB2, receptor accessory protein 5, receptor expression enhancing protein 5, receptor expression-enhancing protein 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction