Learn More
REEP5 Rabbit, anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-14643-100UL
Description
REEP5 (receptor expression-enhancing protein 5), also named as C5orf18, DP1 or TB2, is a membrane protein belonging to DP1/Yop1p protein family. All REEP proteins contain a conserved TB2/DP1, HVA22 domain which is involved in intracellular trafficking and secretion (PMID: 19123125). REEP5 is expressed in circumvallate papillae and testis, and may have the function to influence functional cell surface expression of taste receptors (PMID: 16720576). REEP5 can interact with atlastins (ATL1 and ATL2) which are involved in modulating the structure of ER network (PMID: 19665976).
Specifications
| REEP5 | |
| Polyclonal | |
| CoraLite 488 | |
| 100 μL | |
| 7905 | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |