missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REG1 alpha (REG1A) Mouse anti-Human, Clone: OTI3E8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813961S
This item is not returnable.
View return policy
Description
REG1 alpha (REG1A) Monoclonal antibody specifically detects REG1 alpha (REG1A) in Human samples. It is validated for ELISASpecifications
| REG1 alpha (REG1A) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05451 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI3E8 | |
| Unconjugated | |
| REG1A | |
| ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI | |
| Human recombinant protein fragment corresponding to amino acids 23-166 of human PSP (NP_002900) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction