missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REG1A, Mouse anti-Human, Clone: 3E2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005967-M15.100ug
This item is not returnable.
View return policy
Description
Sequence: MNSGNLVSVLTQAEGAFVASLIKESGTDDFNVWIGLHDPKKNRRWHWSSGSLVSYKSWGIGAPSSVNPGYCVSLTSSTGFQKWKDVPCEDKFSFVCKFKNSpecifications
| REG1A | |
| Monoclonal | |
| Unconjugated | |
| regenerating islet-derived 1 alpha | |
| ICRF/MGC12447/P19/PSP/PSPS/PSPS1/PTP/REG | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 3E2 | |
| In 1x PBS, pH 7.4 | |
| BC005350 | |
| REG1A | |
| REG1A (AAH05350, 67 a.a. ∼ 166 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5967 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido