missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REG1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561038
This item is not returnable.
View return policy
Description
Immunogen sequence: TDDFNVWIGL HDPKKNRRWH WSSGSLVSYK SWG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 76%.
This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication.
Specifications
| REG1A | |
| Polyclonal | |
| Unconjugated | |
| REG1A | |
| ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5967 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05451 | |
| REG1A | |
| Recombinant protein corresponding to Human REG1A. Recombinant protein control fragment (Product #RP-99484). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction