missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg2 Antibody (RM0312-5D45), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12429G
This item is not returnable.
View return policy
Description
Reg2 Monoclonal specifically detects Reg2 in Mouse samples. It is validated for Western Blot.
Specifications
| Reg2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| REG1B | |
| Mouse recombinant Reg2 | |
| 0.1 mL | |
| 5968 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0312-5D45 | |
| Western Blot | |
| regenerating islet-derived 2, RegII | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction