missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REG3A Monoclonal Antibody (02)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531321
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat REG3A.
REG3A encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this protein is observed during pancreatic inflammation and liver carcinogenesis.
Specifications
| REG3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35231 | |
| Reg3a | |
| Recombinant Rat REG3A protein (Met1-Gln174). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| ELISA | |
| 02 | |
| Unconjugated | |
| Reg3a | |
| AV063448; generating islet-derived, mouse homolog 3 alpha; hepatocarcinoma-intestine-pancreas; hepatointestinal pancreatic protein; HIP; HIP/PAP; human proislet peptide; INGAP; islet of Langerhans regenerating protein 3; Lithostathine 3; pancreatic beta cell growth factor; pancreatitis-associated protein 1; Pancreatitis-associated protein 2; PAP; PAP homologous protein; PAP1; Pap2; PAP-H; PapII; PBCGF; proliferation-inducing protein 34; proliferation-inducing protein 42; REG 3; REG 3a; REG 3alpha; Reg IIIalpha; Reg III-alpha; REG3; REG3 alpha; Reg3a; REG3alpha; REG-3alpha; REG-3-alpha; regeneratin; regenerating family member 3 alpha; regenerating islet-derived 3 alpha; regenerating islet-derived protein 3 alpha; regenerating islet-derived protein 3-alpha; Regenerating islet-derived protein 3-alpha 15 kDa form; Regenerating islet-derived protein 3-alpha 16.5 kDa form; Regenerating islet-derived protein III-alpha; regIII; REG-III; RegIII (alpha) | |
| Mouse | |
| Protein A | |
| RUO | |
| 171162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction