missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12268-1-AP-20UL
This item is not returnable.
View return policy
Description
Reg IV is part of the regenerating gene family within the C-type lectin superfamily. This family is involved in liver, pancreatic, gastric and intestinal cell proliferation and differentiation. Reg IV is a 158-amino acid secretory protein implicated in cell regeneration and/or survival with a definite growth stimulating effect on pancreatic beta cells. It is highly expressed in colorectal, gastric, prostate and other types of cancer. Reg IV-positive tumor cells display different phenotypes including mucus-secreting, enterocyte-like, and undifferentiated.Specifications
| REG4 | |
| Polyclonal | |
| Unconjugated | |
| REG4 | |
| GISP, REG 4, REG IV, REG like protein, REG4, RELP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 445583, 67709, 83998 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q68AX7, Q9BYZ8, Q9D8G5 | |
| Reg4 | |
| REG4 Fusion Protein Ag2934 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction