Learn More
Regadenoson, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Regadenoson |
| CAS | 313348-27-5 |
| Molecular Formula | C15 H18 N8 O5 |
| Quantity | 10 mg |
| Synonym | 2-[4-[(Methylamino)carbonyl]-1H-pyrazol-1-yl]adenosine, CVT 3146, Lexiscan, Regadenoson |
| InChI Formula | InChI=1S/C15H18N8O5/c1-17-13(27)6-2-19-23(3-6)15-20-11(16)8-12(21-15)22(5-18-8)14-10(26)9(25)7(4-24)28-14/h2-3,5,7,9-10,14,24-26H,4H2,1H3,(H,17,27)(H2,16,20,21)/t7-,9-,10-,14-/m1/s1 |
| SMILES | CNC(=O)c1cnn(c1)c2nc(N)c3ncn([C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O)c3n2 |
| IUPAC Name | 1-[6-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-2-yl]-N-methylpyrazole-4-carboxamide |
| Molecular Weight (g/mol) | 390.35 |
| Formula Weight | 390.14 |