Learn More
Ismatec REGLO Peristaltic Pump
Peristaltic pump with separate channel control and MS/CA Click-'N-Go Cassettes
Brand:  Ismatec ISM4412
Additional Details : Weight : 2.70000kg
 View more versions of this product
Product Code. 15752977
  • € 3461.00 / Each
Estimated Shipment
10-12-2020
Add to basket
Description
Description
  • Uses 3-stop color-coded tubing.
  • MS/CA Click-'N-Go cassettes.
    • Simple handling
    • Automatic occlusion setting
    • Defined and reproducible occlusion conditions
    • Each channel allows individual tubing selection
  • Operating panel
    • 7-button membrane keypad
    • Digital display showing flow rates and direction for each channel
  • Operating modes
    • Independent flow rates on each channel
    • Independent direction on each channel
    • Independent timing and pause on each channel
    • Pumping by drive speed (rpm)
    • Pumping by flow rate (mL/min)
    • Factory-set flow rates for all available tube sizes
    • Dispensing by volume (mL)
    • Dispensing by time (0.1 sec up to 999 hrs.)
    • Dispensing by volume within a preset time
    • Interval dispensing by volume or time with a pause
    • Setting the number of dispensing cycles
    • Pause time setting (0.1 sec. up to 999 hrs.)
    • Roller backsteps for drip-free dispensing
    • Reversible rotation direction
  • Calibrating function
    • Independent for each channel
    • By flow rate (mL/min.)
    • By dispensing volume (mL)
  • Digital speed setting
  • Closed loop control for pressure independent speed
Specifications
Specifications
Digital speed setting. Closed-loop control for pressure-independent speed.
4
12
205 x 125 x 170 mm
30 w
Digital
3-stop, color-coded tubing
0.0001 to 24 mL/min. per channel
Stepper motor
Closed-loop control for pressure-independent speed.
100/230 VAC 50/60 Hz
2.7 kg
100/230 VAC