missing translation for 'onlineSavingsMsg'
Learn More

Regorafenib Hydrochloride, MedChemExpress

Product Code. p-200098523
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16445638

Brand: Medchem Express HY13308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Regorafenib Hydrochloride (BAY 73-4506 hydrochloride) is a multi-target inhibitor for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1 with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM, respectively.

Regorafenib Hydrochloride (BAY 73-4506 hydrochloride) is a multi-target inhibitor for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1 with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM, respectively.

Specifications

Chemical Name or Material Regorafenib Hydrochloride
CAS 835621-07-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H16Cl2F4N4O3
Quantity 10 mg
Synonym BAY 73-4506 hydrochloride
Solubility Information DMSO : ≥ 5.6 mg/mL (10.78 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClC1=C(C(F)(F)F)C=C(NC(NC2=C(F)C=C(OC3=CC=NC(C(NC)=O)=C3)C=C2)=O)C=C1.Cl
Molecular Weight (g/mol) 519.28
Formula Weight 519.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.