missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Regorafenib monohydrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Regorafenib (BAY 73-4506) monohydrate is an orally active and potent multi-targeted receptor tyrosine kinase inhibitor, with IC50 values of 13/4.2/46, 22, 7, 1.5 and 2.5 nM for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1, respectively. Regorafenib monohydrate shows very robust antitumor and antiangiogenic activity.
Specifications
Specifications
| Chemical Name or Material | Regorafenib monohydrate |
| CAS | 1019206-88-2 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C21H17ClF4N4O4 |
| Quantity | 10 mg |
| Synonym | BAY 73-4506 monohydrate |
| Solubility Information | DMSO : 50 mg/mL (99.83 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC=C(C(C(F)(F)F)=C1)Cl)NC2=CC=C(OC3=CC(C(NC)=O)=NC=C3)C=C2F.O |
| Molecular Weight (g/mol) | 500.83 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction