Learn More
Brady™ Reinforced Polypropylene Beware of Hot Surface Warning Signs
Click to view available options
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | PIC 315-TRI 100-B7527 |
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Description | Reinforced Polypropylene |
| For Use With (Application) | Warning signs Beware of hot surface |
| Height (Metric) | 100 mm |
| Thickness | 0.369 mm |
| Width (Metric) | 100 mm |
| Temperature Range (Metric) | -40°C to 120°C |
| Shape | Triangular |
| Show More |