Learn More
Brady™ Reinforced Polypropylene Escape Route Signs
Click to view available options
Displayed Text:
Directional arrow
Emergency
Emergency Exit
Emergency Stairs Left Pictogram
Emergency Stairs Right Pictogram
Emergency shower
Escape for disabled (right)
Escape route to the Right
Escape route to the left
Output
Rescue ladder
escape (emergency stairs)
Height (Metric):
148 mm
200 mm
250 mm
297 mm
400 mm
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | PIC 359-297X148-B7527 (BRANDED) |
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Description | Reinforced Polypropylene |
| For Use With (Application) | Signs and panels |
| Height (Metric) | 400 mm |
| Thickness | 0.369 mm |
| Width (Metric) | 200 mm |
| Temperature Range (Metric) | -40°C to 120°C |
| Shape | Oblong |
| Show More |