Learn More
Brady™ Reinforced Polypropylene Escape Route Signs
Product Code. 10547044
Change view
Click to view available options
Displayed Text:
Directional arrow
Emergency
Emergency Exit
Emergency Stairs Left Pictogram
Emergency Stairs Right Pictogram
Emergency shower
Escape for disabled (right)
Escape route to the Right
Escape route to the left
Output
Rescue ladder
escape (emergency stairs)
Height (Metric):
148 mm
200 mm
250 mm
297 mm
400 mm
Unit Size:
Each
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text Height (Metric) unitSize
10547044 Emergency Exit 148 mm Each
10765345 Directional arrow 148 mm Each
10547234 Emergency Exit 200 mm Each
10142584 Emergency shower 250 mm Each
10515514 Emergency 297 mm Each
10326204 Emergency 400 mm Each
10355844 Emergency 148 mm Each
10274304 Emergency 200 mm Each
10441844 Emergency Stairs Left Pictogram 297 mm Each
10305954 Emergency Stairs Right Pictogram 297 mm Each
10040854 escape (emergency stairs) 400 mm Each
10152554 Escape for disabled (right) 400 mm Each
10461254 Escape route to the left 297 mm Each
10011444 Escape route to the left 400 mm Each
10537044 Escape route to the Right 297 mm Each
10645295 Escape route to the Right 400 mm Each
10294894 Output 200 mm Each
10162064 Rescue ladder 148 mm Each
10172064 Rescue ladder 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 10547044 Supplier Brady™ Supplier No. 252426
Specifications
Alternate Part Number PIC 363-297X148-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
For Use With (Application) Signs and panels
Height (Metric) 148 mm
Thickness 0.369 mm
Width (Metric) 297 mm
Temperature Range (Metric) -40°C to 120°C
Shape Oblong
Displayed Text Emergency Exit
Product Type Escape Route Sign
Color White on Green