Learn More
Brady™ Reinforced Polypropylene Escape Route Signs
€ 10.15 - € 19.20
Specifications
Material | Polypropylene |
---|---|
Quantity | 1 Ea. |
Description | Reinforced Polypropylene |
Thickness | 0.369 mm |
Temperature Range (Metric) | -40°C to 120°C |
Product Code | Brand | Displayed Text | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Height (Metric) | Price | Quantity & Availability | ||||
10547044
|
Brady™
252426 |
Emergency Exit | 148 mm |
€ 13.40
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10142584
|
Brady™
252414 |
Emergency shower | 250 mm |
€ 13.40
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10515514
|
Brady™
252420 |
Emergency | 297 mm |
€ 13.40
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10326204
|
Brady™
252421 |
Emergency | 400 mm |
€ 19.15
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10355844
|
Brady™
252415 |
Emergency | 148 mm |
€ 19.20
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10274304
|
Brady™
252425 |
Emergency | 200 mm |
€ 19.20
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10441844
|
Brady™
252441 |
Emergency Stairs Left Pictogram | 297 mm |
€ 19.20
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10305954
|
Brady™
252443 |
Emergency Stairs Right Pictogram | 297 mm |
€ 19.20
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10040854
|
Brady™
252442 |
escape (emergency stairs) | 400 mm |
€ 13.40
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10461254
|
Brady™
252417 |
Escape route to the left | 297 mm |
€ 19.20
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10011444
|
Brady™
252418 |
Escape route to the left | 400 mm |
€ 13.40
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10537044
|
Brady™
252419 |
Escape route to the Right | 297 mm |
€ 19.20
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10645295
|
Brady™
252422 |
Escape route to the right | 400 mm |
€ 13.40
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10294894
|
Brady™
252468 |
Output | 200 mm |
€ 10.15
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10152554
|
Brady™
252440 |
Escape for disabled (right) | 400 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10765345
|
Brady™
252403 |
Directional arrow | 148 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10547234
|
Brady™
252427 |
Emergency Exit | 200 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10162064
|
Brady™
252305 |
Rescue ladder | 148 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10172064
|
Brady™
252306 |
Rescue ladder | 200 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Polypropylene | |
Reinforced Polypropylene | |
-40°C to 120°C | |
White on Green |
1 Ea. | |
0.369 mm | |
Escape Route Sign |