Learn More
Brady™ Reinforced Polypropylene Mandatory Hand Protection Signs
Product Code. 11750174
Click to view available options
Color:
White on Blue
Unit Size:
Each
Product Code. 11750174
Brand: Brady™ 222611
Only null left
Specifications
Alternate Part Number PIC 255-DIA 100-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
Diameter (Metric) 100 mm
For Use With (Application) Mandatory Signs Protective Gloves
Thickness 0.369 mm
Temperature Range (Metric) -40°C to 120°C
Shape Round
Displayed Text Protective gloves
Product Type PPE Safety Sign
Color White on Blue