missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Reinforced Polypropylene Mandatory Signs

Brand:  Brady™ 252100

Additional Details : Weight : 0.04300kg

 View more versions of this product

Product Code. 10507234

  • € 13.25 / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Polypropylene
Reinforced Polypropylene
120°C
Round
Mandatory Sign
1 Ea.
200 mm
0.369 mm
Use Eye Protection
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers