Learn More
Brady™ Reinforced Polypropylene Mandatory Signs: For Pedestrians
€ 13.25 - € 32.30
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10623775
|
Brady™
252116 |
315 mm |
€ 32.30
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
10192024
|
Brady™
252115 |
200 mm |
€ 13.25
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||