Learn More
Brady™ Reinforced Polypropylene Parking Prohibited Sign
Product Code. 10663575 Shop All Brady Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10663575 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10663575 Supplier Brady™ Supplier No. 252049
  • ONLINE EXCLUSIVE PRICE
    € 13.60
    € 12.26 / Pack of 1
    Save € 1.34
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use the Reinforced Polypropylene Parking Prohibited Sign for site identification.
  • Brady B-7527 is a surface printed rigid polypropylene material with a slightly textured surface and overlaminated with a gloss clear polyester.
Specifications
Alternate Part Number PIC 225-DIA 200-B7527
Material Polypropylene
Quantity 1 Ea.
Style Indoor/Outdoor Use
Diameter (English) 7.87 in.
Diameter (Metric) 200 mm
For Use With (Application) Facility Identification
Max. Temperature (English) 248°F
Min. Temperature (English) -40°F
Thickness 1.55 mm
Temperature Range (Metric) -40°C to +120°C
Shape Round
Temperature Range (English) -40°F to +248°F
Product Type No Parking Traffic Sign
Color Black/Red on White