Learn More
Brady™ Reinforced Polypropylene Radiation Laser Beam Warning Sign
Click to view available options
Color:
Black on Yellow
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | PIC 309-TRI 100-B7527 |
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Description | Reinforced Polypropylene |
| For Use With (Application) | Used for a wide variety of applications both indoors and outdoors |
| Max. Temperature (Metric) | 120°C |
| Thickness | 0.369 mm |
| Width (Metric) | 100 mm |
| Shape | Triangular |
| Displayed Text | Laser beam warning |
| Show More |