Learn More
Brady™ Reinforced Polypropylene Warning Signs
Brand:  Brady™ 252248
Additional Details : Weight : 0.02660kg
 View more versions of this product
Product Code. 10021734
  • € 9.02 / Each
Estimated Shipment: 05-11-2021
Specifications
ISO 7010
Warning about skid risk
Polypropylene
Used for a wide variety of applications both indoors and outdoors
0.369 mm
200 mm
Black on Yellow
Triangular
Reinforced Polypropylene
200 mm
Warning Sign
-40°C to 120°C