Learn More
Memmert™ Reinforced Stainless Steel Grid
Product Code. 17199624 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17199624
Brand: Memmert™ HPP1060ecoB32550
Only null left
Add to basket
Add to basket
Additional reinforced stainless steel grid for reinforced chamber option
  • Consider max. loading of chamber; electropolished, max. loading 60 kg
Specifications
For Use With (Equipment) HPP1060eco