missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin-3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 168.00 - € 507.00
Specifications
| Antigen | Relaxin-3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626836
|
Novus Biologicals
NBP2-41317-0.025mg |
0.025 mg |
€ 168.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18135338
|
Novus Biologicals
NBP2-41317 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Relaxin-3 Polyclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Relaxin-3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| H3, insl7, relaxin 3, RXN3, ZINS4 | |
| RLN3 | |
| IgG | |
| Affinity Purified | |
| 16 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Neuroscience, Neurotransmission | |
| PBS with 0.02% Sodium Azide | |
| 117579 | |
| Antibody was raised against a 19 amino acid peptide near the center of human RLN3. The immunogen is located within amino acids 80 - 130 of RLN3. Amino Acid Squence: LALTKSPQAFYRGRPSWQG | |
| Primary | |
| RLN3 antibody is human specific. RLN3 antibody is predicted to not cross-react with other members of the Relaxin protein family. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title