missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin R2/LGR8 Antibody (1013903) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB102501JF525
This item is not returnable.
View return policy
Description
Relaxin R2/LGR8 Monoclonal antibody specifically detects Relaxin R2/LGR8 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Relaxin R2/LGR8 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| G protein coupled receptor affecting testicular descent, GPR106G-protein coupled receptor 106, GREATRelaxin family peptide receptor 2, INSL3R, leucine-rich repeat-containing G protein-coupled receptor 8, Leucine-rich repeat-containing G-protein coupled receptor 8, LGR8.1, LGR8G-protein coupled receptor affecting testicular descent, relaxin receptor 2, relaxin/insulin-like family peptide receptor 2, RXFPR2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R2, Met1-Asn416, Accession # Q8WXD0 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 1013903 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 122042 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction