missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelB Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22980UV
This item is not returnable.
View return policy
Description
RelB Polyclonal antibody specifically detects RelB in Human, Mouse samples. It is validated for Western Blot
Specifications
| RelB | |
| Polyclonal | |
| Western Blot | |
| I-Rel, IREL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B | |
| The immunogen recognized by this antibody maps to a region between residue 529 and 579 of human v-rel reticuloendotheliosis viral oncogene homolog B using the numbering given in entry NP_006500.2 (GeneID 5971). | |
| 0.1 mL | |
| Phospho Specific | |
| 5971 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction