missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20122
This item is not returnable.
View return policy
Description
RelB Polyclonal specifically detects RelB in Human samples. It is validated for Western Blot.
Specifications
| RelB | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| IREL, I-REL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B | |
| Rabbit | |
| 62 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (96%), Chicken (84%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000 | |
| Q01201 | |
| RELB | |
| Recombinant protein encompassing a sequence within the center region of human RelB. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5971 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction