missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELM beta, Rat anti-Mouse, Clone: MRB 46L, Novus Biologicals™
Rat Monoclonal Antibody
€ 359.00 - € 484.00
Specifications
| Antigen | RELM beta |
|---|---|
| Clone | MRB 46L |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217616
|
Novus Biologicals
NBP2-80033-50UG |
50 μg |
€ 359.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253638
|
Novus Biologicals
NBP2-80033-100UG |
100 μg |
€ 484.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RELM beta Monoclonal specifically detects RELM beta in Mouse samples. It is validated for Western Blot, ELISA.Specifications
| RELM beta | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Cancer | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| 84666 | |
| Recombinant mouse RELM beta. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| MRB 46L | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
| RETNLB | |
| IgG2a κ | |
| Protein G purified | |
| Recognizes mouse RELM beta. Detects a band of ∽10kDa by Western blot. Does not cross-react with human RELM beta. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title