missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13217
This item is not returnable.
View return policy
Description
REM1 Polyclonal specifically detects REM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| REM1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| GESGD:REM, GTPase GES, GTPase-regulating endothelial cell sprouting, GTP-binding protein REM 1, MGC48669, Rad and Gem-like GTP-binding protein 1, RAS (RAD and GEM)-like GTP-binding, RAS (RAD and GEM)-like GTP-binding 1, REM | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| REM1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QEAKTPLHRRASTPLPLSPRGHQPGRLSTVPSTQSQHPRLGQSASLNPPTQKPSPAPDDWSSESSDSEGSWEALYRVVLLGD | |
| 0.1 mL | |
| Signal Transduction | |
| 28954 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction