missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Remdesivir

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Remdesivir is a viral RNA-dependent RNA polymerase inhibitor (RdRp) and broad spectrum antiviral nucleotide prodrug. Metabolized to active form, GS 441524 (Cat. No. 7227). Exhibits activity against a range of RNA viruses in vitro (EC50 values are 0.019, 0.07, 0.07 - 0.34, 0.086 - 0.14, and 23.15 μM against RSV, SARS-CoV, MERS-CoV, EBOV and SARS-CoV-2, respectively). Exhibits antiviral efficacy in animal models of MERS-CoV and EBOV infection.
Specifications
Specifications
| Chemical Name or Material | 2-Ethylbutyl (2 S)-2-[[-[[(2 R,3 S,4 R,5 R)-5-(4-aminopyrrolo(2,1-f)(1,2,4)triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl]methoxy]phenoxyphosphoryl]amino]propanoate |
| CAS | 1809249-37-3 |
| Quantity | 50 mg |
| Target | RNA/DNA Polymerase Inhibitors |
| Molecular Formula | C27H35N6O8P |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction