Learn More
Thermo Scientific™ Remel™ Gram Decolorizer
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
Brand: Thermo Scientific™ R40075
Additional Details : Weight : 4.38300kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
Specifications
3.785 L | |
Decolorizer | |
Each | |
CE Marked |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.